(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥90.0%

Supplier: TCI America

Synonyms: (R,R)-(-)-DDB, (R,R)-(-)-2,3-Dimethoxy-N,N,N',N'-tetramethyl-1,4-butanediamine

D2395-1G D2395-5G
TCD2395-001GEA 293.88 CAD
TCD2395-001G TCD2395-005G
(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥90.0%
(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane

CAS Number: 26549-22-4
MDL Number: MFCD00008331
Molecular Formula: C10H24N2O2
Molecular Weight: 204.31
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: -10.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-(-)-DDB, (R,R)-(-)-2,3-Dimethoxy-N,N,N',N'-tetramethyl-1,4-butanediamine

Formula: C₁₀H₂₄N₂O₂
MW: 204.31 g/mol
Boiling Pt: 64 °C
Density: 0.90 (20 °C)
MDL Number: MFCD00008331
CAS Number: 26549-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1e2c23e92764f9ca25e07a7f4f45ab68ff96f44&vid=c1e2c23e92764f9ca25e07a7f4f45ab68ff96f44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR