Trimethylene di(thiotosylate) ≥95.0% (by HPLC)

Supplier: TCI America

D2390-25G D2390-5G
TCD2390-25GEA 312.84 CAD
TCD2390-25G TCD2390-005G
Trimethylene di(thiotosylate) ≥95.0% (by HPLC)
Trimethylene di(thiotosylate)

[Protecting Reagent for Active Methylene]
CAS Number: 3866-79-3
MDL Number: MFCD00014918
Molecular Formula: C17H20O4S4
Molecular Weight: 416.58
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 66


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 1,3-Di(p-tosylthio)propane, 1,3-Propanediyl Di(thiotosylate), Trimethylene Bis(p-toluenethiosulfonate)

Formula: C₁₇H₂₀O₄S₄
Melting Pt: 66 °C
MDL Number: MFCD00014918
CAS Number: 3866-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4df5adc085aebef12d559c61427efd6d0b5b79db&vid=4df5adc085aebef12d559c61427efd6d0b5b79db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR