3,4-Diaminothiophene dihydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Thiophene-3,4-diamine dihydrochloride

D2378-1G D2378-100MG
TCD2378-1GEA 208.56 CAD
TCD2378-1G TCD2378-100MG
3,4-Diaminothiophene dihydrochloride ≥98.0% (by HPLC)
3,4-Diaminothiophene dihydrochloride

CAS Number: 90069-81-1
MDL Number: MFCD00671511
Molecular Formula: C4H6N2S
Molecular Weight: 187.08
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₄H₆N₂S
MW: 187.08 g/mol
MDL Number: MFCD00671511
CAS Number: 90069-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71ce0c03a92f42b6d3f9bd8d5bb1fede2604ad73&vid=71ce0c03a92f42b6d3f9bd8d5bb1fede2604ad73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR