2,4-Diethylthioxanthen-9-one ≥98.0%

Supplier: TCI America

D2375-250G D2375-25G
TCD2375-250GEA 504.02 CAD
TCD2375-250G TCD2375-025G
2,4-Diethylthioxanthen-9-one ≥98.0%
2,4-Diethylthioxanthen-9-one

CAS Number: 82799-44-8
MDL Number: MFCD00082910
Molecular Formula: C17H16OS
Molecular Weight: 268.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₁₇H₁₆OS
Melting Pt: 70 °C
MDL Number: MFCD00082910
CAS Number: 82799-44-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86c180a86916b217c4b2bb7f7ba5a63e33d5a545&vid=86c180a86916b217c4b2bb7f7ba5a63e33d5a545&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR