5,5-Dimethyl-1-pyrroline-N-oxide 97,0

Supplier: TCI America

Synonyms: DMPO, 5,5-Dimethyl-1-pyrroline N-oxide

D2362-5G D2362-1G
TCD2362-5GEA 752.08 CAD
TCD2362-5G TCD2362-001G
5,5-Dimethyl-1-pyrroline-N-oxide 97,0
5,5-Dimethyl-1-pyrroline-N-oxide

CAS Number: 3317-61-1
MDL Number: MFCD00005279
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Flash Point (°C): 95
Freezing point (°C): 30
Storage Temperature: <0°C


Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: DMPO

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Flash Pt: 95 °C
MDL Number: MFCD00005279
CAS Number: 3317-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b1df8b59e8b7db5bc78a34327e28ae90c63abd0&vid=1b1df8b59e8b7db5bc78a34327e28ae90c63abd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR