Diphenoxymethane ≥98.0%
Supplier: TCI America
D2352-25G
D2352-5G
TCD2352-025GEA
421.86
CAD
TCD2352-025G
TCD2352-5G
Diphenoxymethane ≥98.0%
Diphenoxymethane
CAS Number: 4442-41-5
MDL Number: MFCD00191642
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Freezing point (°C): 15
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: Formaldehyde Diphenyl Acetal, Methylene Glycol Diphenyl Ether
Formula:
C₁₃H₁₂O₂ Boiling Pt: 122 °C Density: 1.12 (20 °C) |
MDL Number:
MFCD00191642 CAS Number: 4442-41-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...