4-Methyl-N-phenyl-N-(p-tolyl)aniline ≥98.0%
Supplier: TCI America
D2347-25G
D2347-1G
D2347-5G
TCD2347-25GEA
285.98
CAD
TCD2347-25G
TCD2347-001G
TCD2347-005G
4-Methyl-N-phenyl-N-(p-tolyl)aniline ≥98.0%
4-Methyl-N-phenyl-N-(p-tolyl)aniline
CAS Number: 20440-95-3
MDL Number: MFCD00145004
Molecular Formula: C20H19N
Molecular Weight: 273.38
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110
Other Product Names / Synonyms: Bis(4-methylphenyl)phenylamine, N,N-Di-p-tolylaniline, Phenyl(di-p-tolyl)amine
Formula:
C₂₀H₁₉N Melting Pt: 110 °C |
MDL Number:
MFCD00145004 CAS Number: 20440-95-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...