N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-12, SB-12) ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Danger
|
Synonyms:
sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulfonate, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate, sulfobetaine-12
CAS Number: 14933-08-5
MDL Number: MFCD00036909
Molecular Formula: C17H37NO3S
Molecular Weight: 335.55
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 244
Flash Point (°C): 110
Product Categories: Materials Chemistry, Amphoteric Surfactants
Other Product Names / Synonyms: 3-(Dodecyldimethylammonio)propane-1-sulfonate, Lauryldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, Lauryl Sulfobetaine, 3-(Lauryldimethylammonio)propane-1-sulfonate
Formula:
C₁₇H₃₇NO₃S MW: 335.55 g/mol Melting Pt: 244 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036909 CAS Number: 14933-08-5 EINECS: 239-002-3 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...