4,4'-Dimethoxyoctafluorobiphenyl ≥98.0%

Supplier: TCI America

Synonyms: Octafluoro-4,4'-dimethoxybiphenyl

D2330-1G D2330-5G
TCD2330-1GEA 60.04 CAD
TCD2330-1G TCD2330-005G
4,4'-Dimethoxyoctafluorobiphenyl ≥98.0%
4,4'-Dimethoxyoctafluorobiphenyl

CAS Number: 2200-71-7
MDL Number: MFCD00039802
Molecular Formula: C14H6F8O2
Molecular Weight: 358.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: Octafluoro-4,4'-dimethoxybiphenyl

Formula: C₁₄H₆F₈O₂
MW: 358.19 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00039802
CAS Number: 2200-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=661ae66515b15c96c0c72ee7322bba88496729c8&vid=661ae66515b15c96c0c72ee7322bba88496729c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR