(2,6-Dichlorophenyl)acetic acid methyl ester ≥98.0%
Supplier: TCI America
D2328-25G
D2328-5G
TCD2328-25GEA
368.61
CAD
TCD2328-25G
TCD2328-005G
(2,6-Dichlorophenyl)acetic acid methyl ester ≥98.0%
(2,6-Dichlorophenyl)acetic acid methyl ester
CAS Number: 54551-83-6
MDL Number: MFCD00191640
Molecular Formula: C9H8Cl2O2
Molecular Weight: 219.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.33
Other Product Names / Synonyms: 2,6-Dichlorophenylacetic Acid Methyl Ester
Formula:
(C₆H₃Cl₂)CH₂COOCH₃ Density: 1.33 (20 °C) |
MDL Number:
MFCD00191640 CAS Number: 54551-83-6 EINECS: 259-221-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...