tert-Butyl-2-((diphenylmethylene)amino)acetate ≥98.0%

Supplier: TCI America

Synonyms: N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate

D2322-5G D2322-1G D2322-25G
TCD2322-5GEA 56.26 CAD
TCD2322-5G TCD2322-001G TCD2322-25G
tert-Butyl-2-((diphenylmethylene)amino)acetate ≥98.0%
tert-Butyl-2-((diphenylmethylene)amino)acetate

CAS Number: 81477-94-3
MDL Number: MFCD00134280
Molecular Formula: C19H21NO2
Molecular Weight: 295.38
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 116


Product Categories: Synthetic Organic Chemistry, Glycine Equivalents
Other Product Names / Synonyms: tert-Butyl (Diphenylmethyleneamino)acetate, (Diphenylmethyleneamino)acetic Acid tert-Butyl Ester

Formula: C₁₉H₂₁NO₂
MW: 295.382 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00134280
CAS Number: 81477-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9366e952120a3049a8f102427a530339da1597e8&vid=9366e952120a3049a8f102427a530339da1597e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR