N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether ≥98.0%

Supplier: TCI America

Synonyms: Mercury ionophore I, N,N'-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane

D2321-5G D2321-1G
TCD2321-5GEA 851.62 CAD
TCD2321-5G TCD2321-001G
N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether ≥98.0%
N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether

CAS Number: 69703-25-9
MDL Number: MFCD00010831
Molecular Formula: C26H38N2O4
Molecular Weight: 442.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 84


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: N,N'-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane

Formula: C₂₆H₃₈N₂O₄
MW: 442.60 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00010831
CAS Number: 69703-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d392275c94064fd0aed2ff3e67376943f9281a5e&vid=d392275c94064fd0aed2ff3e67376943f9281a5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR