Dimethoxymethylphenylsilane ≥98.0%

Supplier: TCI America

Synonyms: Methylphenyldimethoxysilane, Dimethoxy(methyl)(phenyl)silane

D2319-100ML D2319-25ML
TCD2319-100MLEA 135.88 CAD
TCD2319-100ML TCD2319-025ML
Dimethoxymethylphenylsilane ≥98.0%
Dimethoxymethylphenylsilane

CAS Number: 3027-21-2
MDL Number: MFCD00008343
Molecular Formula: C9H14O2Si
Molecular Weight: 182.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 82
Specific Gravity (20/20): 1.01

Formula: C₉H₁₄O₂Si
MW: 182.29 g/mol
Boiling Pt: 199 °C
Density: 1.01 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00008343
CAS Number: 3027-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03862773e7a6f32a65407a5be3181ab6b7216c33&vid=03862773e7a6f32a65407a5be3181ab6b7216c33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR