5,6-Diacetoxyindole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D2317-1G
D2317-5G
TCD2317-1GEA
188.49
CAD
TCD2317-1G
TCD2317-005G
5,6-Diacetoxyindole ≥98.0% (by HPLC, total nitrogen)
5,6-Diacetoxyindole
CAS Number: 15069-79-1
MDL Number: MFCD00152051
Molecular Formula: C12H11NO4
Molecular Weight: 233.22
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 138
Formula:
C₁₂H₁₁NO₄ Melting Pt: 138 °C |
MDL Number:
MFCD00152051 CAS Number: 15069-79-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...