2-Amino-6-chloropyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-4-chloro-6-pyrimidinol, 2-Amino-4-chloro-6-hydroxypyrimidine

A1109-25G A1109-5G
TCA1109-025GEA 471.16 CAD
TCA1109-025G TCA1109-005G
2-Amino-6-chloropyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chloropyrimidin-4(3H)-one

CAS Number: 1194-21-4
MDL Number: MFCD00006093
Molecular Formula: C4H4ClN3O
Molecular Weight: 145.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-4-chloro-6-pyrimidinol

Formula: C₄H₄ClN₃O
MW: 145.5500 g/mol
MDL Number: MFCD00006093
CAS Number: 1194-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d738f06947c90eacbd756115f0ddc56806a78c1&vid=9d738f06947c90eacbd756115f0ddc56806a78c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR