2-Dodecylphenol ≥98.0%

Supplier: TCI America

D2302-10G D2302-25G
TCD2302-010GEA 421.86 CAD
TCD2302-010G TCD2302-25G
2-Dodecylphenol ≥98.0%
2-Dodecylphenol

CAS Number: 5284-29-7
MDL Number: MFCD00020106
Molecular Formula: C18H30O
Molecular Weight: 262.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46


Other Product Names / Synonyms: 2-Laurylphenol

Formula: C₁₈H₃₀O
Melting Pt: 46 °C
MDL Number: MFCD00020106
CAS Number: 5284-29-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a6092128970a1319441857339e5c08a624793a5&vid=1a6092128970a1319441857339e5c08a624793a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR