Dipyridamole

Supplier: TCI America

Synonyms: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetrakis(ethan-1-ol), 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

D2274-25G D2274-5G
TCD2274-25GEA 210.5 CAD
TCD2274-25G TCD2274-005G
Dipyridamole
Dipyridamole

CAS Number: 58-32-2
MDL Number: MFCD00010555
Molecular Formula: C24H40N8O4
Molecular Weight: 504.64
Form: Crystal
Color: Pale Yellow
Melting point (°C): 168


Other Product Names / Synonyms: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

Formula: C₂₄H₄₀N₈O₄
MW: 504.64 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00010555
CAS Number: 58-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2da375d3cbc91ce1e02f3f88da5c3e08de26e2&vid=ee2da375d3cbc91ce1e02f3f88da5c3e08de26e2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR