3,5-Dibenzyloxybenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,5-Bis(benzyloxy)benzoic acid

D2267-5G D2267-25G
TCD2267-005GEA 157.05 CAD
TCD2267-005G TCD2267-025G
3,5-Dibenzyloxybenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Dibenzyloxybenzoic acid

CAS Number: 28917-43-3
MDL Number: MFCD00191341
Molecular Formula: C21H18O4
Molecular Weight: 334.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 214


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(benzyloxy)benzoic Acid

Formula: C₂₁H₁₈O₄
MW: 334.37 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00191341
CAS Number: 28917-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=586cd5548842468f78b66be054ed43f3ed0e91ec&vid=586cd5548842468f78b66be054ed43f3ed0e91ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR