1,2:5,6-Di-O-isopropylidene-α-D-allofuranose ≥98.0%
Supplier: TCI America
D2265-5G
D2265-1G
TCD2265-5GEA
401.32
CAD
TCD2265-5G
TCD2265-001G
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose ≥98.0%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
CAS Number: 2595-05-3
MDL Number: MFCD00135634
Molecular Formula: C12H20O6
Molecular Weight: 260.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 75
Specific rotation [a]20/D: 36.5 deg (C=1, CHCl3)
Storage Temperature: <0°C
Formula:
C₁₂H₂₀O₆ Melting Pt: 75 °C |
MDL Number:
MFCD00135634 CAS Number: 2595-05-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...