4-Dimethylamino-4'-nitrobiphenyl ≥98.0% (by HPLC)

Supplier: TCI America

D2263-1G D2263-5G
TCD2263-001GEA 556.16 CAD
TCD2263-001G TCD2263-005G
4-Dimethylamino-4'-nitrobiphenyl ≥98.0% (by HPLC)
4-Dimethylamino-4'-nitrobiphenyl

CAS Number: 2143-87-5
MDL Number: MFCD00191632
Molecular Formula: C14H14N2O2
Molecular Weight: 242.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 246

Formula: C₁₄H₁₄N₂O₂
Melting Pt: 246 °C
MDL Number: MFCD00191632
CAS Number: 2143-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3b2076cf81bd3adb7ef8f051e493d0ffc45fa52&vid=a3b2076cf81bd3adb7ef8f051e493d0ffc45fa52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR