Dichlorotitanium diisopropoxide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D2257-5G D2257-25G
TCD2257-005GEA 178.54 CAD
TCD2257-005G TCD2257-25G
Dichlorotitanium diisopropoxide ≥95.0% (by titrimetric analysis)
Dichlorotitanium diisopropoxide

CAS Number: 762-99-2
MDL Number: MFCD00191416
Molecular Formula: C6H14Cl2O2Ti
Molecular Weight: 236.94
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Flash Point (°C): 113
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Dichloro(diisopropoxy)titanium

Formula: C₆H₁₄Cl₂O₂Ti
Flash Pt: 113 °C
MDL Number: MFCD00191416
CAS Number: 762-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d382986ee3bad51077247543cc2ead8a7179d7&vid=66d382986ee3bad51077247543cc2ead8a7179d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR