2,5-Dimethoxy-1,4-benzoquinone ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Dimethoxy-p-quinone

D2250-25G D2250-5G
TCD2250-25GEA 1074.4 CAD
TCD2250-25G TCD2250-005G
2,5-Dimethoxy-1,4-benzoquinone ≥98.0%
2,5-Dimethoxy-1,4-benzoquinone

CAS Number: 3117-03-1
MDL Number: MFCD00189386
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2,5-Dimethoxy-p-quinone

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00189386
CAS Number: 3117-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68bd1abcedc5508ff15846aa01edf9651d331006&vid=68bd1abcedc5508ff15846aa01edf9651d331006&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR