Acrylic acid ≥99.0% stabilised for synthesis

Supplier: TCI America
Danger

Synonyms: Prop-2-enoic acid

A0141-25G A0141-500G
TCA0141-025GEA 56.55 CAD
TCA0141-025G TCA0141-500G
Acrylic acid ≥99.0% stabilised for synthesis
Acrylic acid

(stabilized with MEHQ)
CAS Number: 79-10-7
MDL Number: MFCD00004367
Molecular Formula: C3H4O2
Molecular Weight: 72.06
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 51
Freezing point (°C): 13
Specific Gravity (20/20): 1.05
Lambda max.: 231 nm

Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 141 °C
Melting Pt: 13
Density: 1.05 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=288a99c09cf862a61728546ef9b0110820491d96&vid=288a99c09cf862a61728546ef9b0110820491d96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR