2,5-Dibromo-1,4-benzoquinone ≥98.0%
Supplier: TCI America
D2249-5G
D2249-1G
TCD2249-005GEA
1161.3
CAD
TCD2249-005G
TCD2249-001G
2,5-Dibromo-1,4-benzoquinone ≥98.0%
2,5-Dibromo-1,4-benzoquinone
CAS Number: 1633-14-3
MDL Number: MFCD00156859
Molecular Formula: C6H2Br2O2
Molecular Weight: 265.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 193
Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2,5-Dibromo-p-quinone
Formula:
C₆H₂Br₂O₂ Melting Pt: 193 °C |
MDL Number:
MFCD00156859 CAS Number: 1633-14-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...