2,8-Dibromdibenzothiophene ≥96.0%
Supplier: TCI America
D2245-5G
D2245-1G
D2245-25G
TCD2245-5GEA
175.17
CAD
TCD2245-5G
TCD2245-001G
TCD2245-25G
2,8-Dibromdibenzothiophene ≥96.0%
2,8-Dibromdibenzothiophene
CAS Number: 31574-87-5
MDL Number: MFCD00092755
Molecular Formula: C12H6Br2S
Molecular Weight: 342.05
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 226
Formula:
C₁₂H₆Br₂S Melting Pt: 226 °C |
MDL Number:
MFCD00092755 CAS Number: 31574-87-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...