(-)-1,4-Di-O-benzyl-L-threitol ≥98.0%

Supplier: TCI America

D2240-1G D2240-5G
TCD2240-001GEA 309.68 CAD
TCD2240-001G 76219-002
(-)-1,4-Di-O-benzyl-L-threitol ≥98.0%
(-)-1,4-Di-O-benzyl-L-threitol

CAS Number: 17401-06-8
MDL Number: MFCD00077723
Molecular Formula: C18H22O4
Molecular Weight: 302.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56
Specific rotation [a]20/D: -6.3 deg (C=5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others)
Other Product Names / Synonyms: (-)-(2S,3S)-1,4-Bis(benzyloxy)-2,3-butanediol

Formula: C₁₈H₂₂O₄
Melting Pt: 56 °C
MDL Number: MFCD00077723
CAS Number: 17401-06-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1efc86fc9130314661cc78ac462484fd811ecb19&vid=1efc86fc9130314661cc78ac462484fd811ecb19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR