4-Dodecyloxyphthalonitrile ≥99.0%

Supplier: TCI America

D2237-1G D2237-5G
TCD2237-001GEA 91.61 CAD
TCD2237-001G TCD2237-005G
4-Dodecyloxyphthalonitrile ≥99.0%
4-Dodecyloxyphthalonitrile

CAS Number: 161082-75-3
MDL Number: MFCD00191413
Molecular Formula: C20H28N2O
Molecular Weight: 312.46
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 62


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-4-dodecyloxybenzene

Formula: C₂₀H₂₈N₂O
Melting Pt: 62 °C
MDL Number: MFCD00191413
CAS Number: 161082-75-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbe50e45865a32ab9744b3de2617bd70aaffd6e6&vid=fbe50e45865a32ab9744b3de2617bd70aaffd6e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR