2,4-Diamino-6-dimethylamino-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: N2,N2-Dimethyl-1,3,5-triazine-2,4,6-triamine, Metformin Related Compound C, N2,N2-Dimethylmelamine

D2236-5G D2236-1G
TCD2236-005GEA 748.92 CAD
TCD2236-005G 76219-000
2,4-Diamino-6-dimethylamino-1,3,5-triazine ≥98.0%
2,4-Diamino-6-dimethylamino-1,3,5-triazine

CAS Number: 1985-46-2
MDL Number: MFCD00191338
Molecular Formula: C5H10N6
Molecular Weight: 154.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N2,N2-Dimethylmelamine

Formula: C₅H₁₀N₆
MW: 154.18 g/mol
MDL Number: MFCD00191338
CAS Number: 1985-46-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6587406c562085520010a0be0eeec3b9bcf5ec15&vid=6587406c562085520010a0be0eeec3b9bcf5ec15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR