9-Decenyl acetate ≥97.0%

Supplier: TCI America

A1093-25ML A1093-5ML
TCA1093-025MLEA 486.99 CAD
TCA1093-025ML TCA1093-5ML
9-Decenyl acetate ≥97.0%
9-Decenyl acetate

CAS Number: 50816-18-7
MDL Number: MFCD00036512
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Acetic Acid 9-Decenyl Ester, 10-Acetoxy-1-decene

Formula: C₁₂H₂₂O₂
Boiling Pt: 246 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00036512
CAS Number: 50816-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cbf48155b6c3f4379e0d2631bb17c3a9f6e3708&vid=8cbf48155b6c3f4379e0d2631bb17c3a9f6e3708&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR