2,4-Diamino-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: 1,3,5-Triazine-2,4-diamine

D2227-5G D2227-1G
TCD2227-5GEA 703.1 CAD
TCD2227-5G TCD2227-001G
2,4-Diamino-1,3,5-triazine ≥98.0%
2,4-Diamino-1,3,5-triazine

CAS Number: 504-08-5
MDL Number: MFCD00014598
Molecular Formula: C3H5N5
Molecular Weight: 111.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 325

Formula: C₃H₅N₅
MW: 111.11 g/mol
Melting Pt: 325 °C
MDL Number: MFCD00014598
CAS Number: 504-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40ebd1033fe900bc1a3f5d032bbfe60dda444216&vid=40ebd1033fe900bc1a3f5d032bbfe60dda444216&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR