3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole ≥96.0%

Supplier: TCI America

Synonyms: Ethyl 3,4-Diethyl-5-methyl-2-pyrrolecarboxylate, 3,4-Diethyl-5-methyl-2-pyrrolecarboxylic Acid Ethyl Ester

D2217-5G D2217-1G
TCD2217-5GEA 674.34 CAD
TCD2217-5G TCD2217-001G
3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole ≥96.0%
3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole

CAS Number: 16200-50-3
MDL Number: MFCD00075089
Molecular Formula: C12H19NO2
Molecular Weight: 209.29
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 76


Other Product Names / Synonyms: 3,4-Diethyl-5-methyl-2-pyrrolecarboxylic Acid Ethyl Ester, Ethyl 3,4-Diethyl-5-methyl-2-pyrrolecarboxylate

Formula: C₁₂H₁₉NO₂
MW: 209.29 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00075089
CAS Number: 16200-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2a4dfa7cc68735919bb62bf845cde0c57c411ac&vid=d2a4dfa7cc68735919bb62bf845cde0c57c411ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR