2,11-Dithia[3.3]paracyclophane ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
3,10-Dithiatricyclo[10.2.2.2(5,8)]octadeca-5,7,12,14,15,17-hexaene
D2210-100MG
D2210-500MG
TCD2210-100MGEA
74.42
CAD
TCD2210-100MG
TCD2210-500MG
2,11-Dithia[3.3]paracyclophane ≥98.0% (by HPLC)
2,11-Dithia[3.3]paracyclophane
CAS Number: 28667-63-2
MDL Number: MFCD00191410
Molecular Formula: C16H16S2
Molecular Weight: 272.42
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 222
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 3,10-Dithiatricyclo[10.2.2.2(5,8)]octadeca-5,7,12,14,15,17-hexaene
Formula:
C₁₆H₁₆S₂ MW: 272.42 g/mol Melting Pt: 222 °C |
MDL Number:
MFCD00191410 CAS Number: 28667-63-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...