meso-2,3-Diphenylsuccinic acid
Supplier: TCI America
Synonyms:
meso-2,3-Diphenylbutanedioic Acid
D2209-5G
D2209-1G
TCD2209-005GEA
595.03
CAD
TCD2209-005G
TCD2209-001G
meso-2,3-Diphenylsuccinic acid
meso-2,3-Diphenylsuccinic acid
CAS Number: 1225-13-4
MDL Number: MFCD00198082
Molecular Formula: C16H14O4
Molecular Weight: 270.28
Form: Crystal
Color: White
Other Product Names / Synonyms: meso-2,3-Diphenylbutanedioic Acid
Formula:
C₁₆H₁₄O₄ MW: 270.28 g/mol |
MDL Number:
MFCD00198082 CAS Number: 1225-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...