2,5-Dimethylterephthalic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D2208-5G D2208-25G
TCD2208-005GEA 489.8 CAD
TCD2208-005G TCD2208-25G
2,5-Dimethylterephthalic acid ≥97.0% (by GC, titration analysis)
2,5-Dimethylterephthalic acid

CAS Number: 6051-66-7
MDL Number: MFCD00020292
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00020292
CAS Number: 6051-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f97971845cb9a4b0a98248c3669f4a457506f839&vid=f97971845cb9a4b0a98248c3669f4a457506f839&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR