3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥98.0%

Supplier: TCI America

Synonyms: Diisopropyl Squarate, 3,4-Diisopropoxycyclobut-3-ene-1,2-dione

D2207-25G D2207-5G
TCD2207-025GEA 703.1 CAD
TCD2207-025G TCD2207-005G
3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥98.0%
3,4-Diisopropoxy-3-cyclobutene-1,2-dione

CAS Number: 61699-62-5
MDL Number: MFCD00075125
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 113


Other Product Names / Synonyms: Diisopropyl Squarate

Formula: C₁₀H₁₄O₄
Melting Pt: 46 °C
Flash Pt: 113 °C
MDL Number: MFCD00075125
CAS Number: 61699-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84287e972c33595640adcd6570ff4a5401e8957b&vid=84287e972c33595640adcd6570ff4a5401e8957b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR