4,4'-Dimethoxybenzhydrylamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D2202-25G D2202-1G D2202-5G
TCD2202-25GEA 617.78 CAD
TCD2202-25G TCD2202-001G TCD2202-005G
4,4'-Dimethoxybenzhydrylamine ≥98.0% (by HPLC, titration analysis)
4,4'-Dimethoxybenzhydrylamine

CAS Number: 19293-62-0
MDL Number: MFCD00191409
Molecular Formula: C15H17NO2
Molecular Weight: 243.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 62


Other Product Names / Synonyms: 1,1-Di(p-anisyl)methylamine

Formula: C₁₅H₁₇NO₂
Melting Pt: 62 °C
MDL Number: MFCD00191409
CAS Number: 19293-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84040bdaf24ce0ed14c97b3bf13cd2288f657734&vid=84040bdaf24ce0ed14c97b3bf13cd2288f657734&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR