4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

D2192-25G D2192-5G
TCD2192-025GEA 257.86 CAD
TCD2192-025G TCD2192-5G
4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride ≥97.0% (by HPLC, titration analysis)
4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride

CAS Number: 13912-65-7
MDL Number: MFCD00030105
Molecular Formula: C16H12O6
Molecular Weight: 300.27
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 204


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4-(2,5-Dioxotetrahydrofuran-3-yl)-tetralin-1,2-dicarboxylic Anhydride

Formula: C₁₆H₁₂O₆
Melting Pt: 204 °C
MDL Number: MFCD00030105
CAS Number: 13912-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1ec445804d344a229fc3f39b09c5f6d480ed6c5&vid=b1ec445804d344a229fc3f39b09c5f6d480ed6c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR