6,8-Dimethoxy-4-methylquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2190-5G D2190-1G
TCD2190-5GEA 1046.28 CAD
TCD2190-5G TCD2190-001G
6,8-Dimethoxy-4-methylquinoline ≥98.0% (by GC, titration analysis)
6,8-Dimethoxy-4-methylquinoline

CAS Number: 51049-14-0
MDL Number: MFCD00191629
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 65

Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 65 °C
MDL Number: MFCD00191629
CAS Number: 51049-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfe11009e851f22bfef98fe56bd52dab23e72f23&vid=cfe11009e851f22bfef98fe56bd52dab23e72f23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR