2',3'-Dihydroxy-4'-methoxyacetophenone hydrate ≥98.0% (by HPLC)

Supplier: TCI America

D2187-5G D2187-25G
TCD2187-005GEA 121.66 CAD
TCD2187-005G TCD2187-025G
2',3'-Dihydroxy-4'-methoxyacetophenone hydrate ≥98.0% (by HPLC)
2',3'-Dihydroxy-4'-methoxyacetophenone hydrate

CAS Number: 708-53-2
MDL Number: MFCD00180772
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: Gallacetophenone-4'-methyl Ether

Formula: C₉H₁₀O₄·H₂O
Melting Pt: 133 °C
MDL Number: MFCD00180772
CAS Number: 708-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73cc77f0032ab09d77ecb1687ba91b1864a91e67&vid=1efbc093defb9a826ff8796aeee3c61f4fe4de75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR