2,5-Dimethyl-p-phenylenediamine ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
2,5-Diamino-p-xylene, 2,5-Dimethyl-1,4-phenylenediamine, 1,4-Diamino-2,5-dimethylbenzene, 2,5-Dimethylbenzene-1,4-diamine
D2183-25G
D2183-100G
TCD2183-025GEA
166.69
CAD
TCD2183-025G
TCD2183-100G
2,5-Dimethyl-p-phenylenediamine ≥98.0% for synthesis
2,5-Dimethyl-p-phenylenediamine
CAS Number: 6393-01-7
MDL Number: MFCD00052675
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2,5-Diamino-p-xylene
Formula:
C₈H₁₂N₂ MW: 136.2 g/mol Melting Pt: 148 °C |
MDL Number:
MFCD00052675 CAS Number: 6393-01-7 EINECS: 229-003-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...