1,5-Dimercaptonaphthalene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2182-1G D2182-5G
TCD2182-001GEA 258.87 CAD
TCD2182-001G TCD2182-5G
1,5-Dimercaptonaphthalene ≥98.0% (by GC, titration analysis)
1,5-Dimercaptonaphthalene

CAS Number: 5325-88-2
MDL Number: MFCD00068909
Molecular Formula: C10H8S2
Molecular Weight: 192.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 121


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,5-Naphthalenedithiol

Formula: C₁₀H₈S₂
Melting Pt: 121 °C
MDL Number: MFCD00068909
CAS Number: 5325-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f756bc22424e50bf82ca8148a4f2416bbdcecc75&vid=f756bc22424e50bf82ca8148a4f2416bbdcecc75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR