3-Amino-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A1067-5G A1067-25G
TCA1067-5GEA 69.52 CAD
TCA1067-5G TCA1067-025G
3-Amino-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)
3-Amino-N-methylbenzamide

CAS Number: 25900-61-2
MDL Number: MFCD00070630
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 119


Other Product Names / Synonyms: 3-Aminobenzoylmethylamide

Formula: C₈H₁₀N₂O
Melting Pt: 119 °C
MDL Number: MFCD00070630
CAS Number: 25900-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54bd0b8f040077fa6beb17758b21ab933d4288cc&vid=54bd0b8f040077fa6beb17758b21ab933d4288cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR