2-Phenyl-2-oxazoline ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenyl(2-oxazoline), (4,5-Dihydro-2-oxazolyl)benzene, 4,5-Dihydro-2-phenyloxazole

D2155-5G D2155-25G
TCD2155-5GEA 150.1 CAD
TCD2155-5G TCD2155-025G
2-Phenyl-2-oxazoline ≥98.0%
2-Phenyl-2-oxazoline

CAS Number: 7127-19-7
MDL Number: MFCD00075541
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 243
Flash Point (°C): 113
Freezing point (°C): 28
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (4,5-Dihydro-2-oxazolyl)benzene, 2-Phenyl(2-oxazoline)

Formula: C₉H₉NO
Boiling Pt: 243 °C
Flash Pt: 113 °C
MDL Number: MFCD00075541
CAS Number: 7127-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745478b78cca650fbfb0474ee94689297441f4cd&vid=745478b78cca650fbfb0474ee94689297441f4cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR