2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran ≥98.0%

Supplier: TCI America

D2154-25G D2154-100G
TCD2154-025GEA 67.94 CAD
TCD2154-025G 76218-998
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran ≥98.0%
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran

CAS Number: 1563-38-8
MDL Number: MFCD00075382
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 113
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 2,2-Dimethyl-7-hydroxycoumaran

Formula: C₁₀H₁₂O₂
Boiling Pt: 95 °C
Density: 1.11 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00075382
CAS Number: 1563-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66047f7c2cdd4ac860c3eefe1f8df8cb9a496bdf&vid=66047f7c2cdd4ac860c3eefe1f8df8cb9a496bdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR