ɑ,ɑ'-Dianilino-p-xylene ≥98.0%
Supplier: TCI America
D2143-1G
D2143-5G
TCD2143-1GEA
191.18
CAD
TCD2143-1G
TCD2143-005G
ɑ,ɑ'-Dianilino-p-xylene ≥98.0%
alpha,alpha'-Dianilino-p-xylene
CAS Number: 13170-62-2
MDL Number: MFCD00143578
Molecular Formula: C20H20N2
Molecular Weight: 288.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127
Formula:
C₂₀H₂₀N₂ Melting Pt: 127 °C |
MDL Number:
MFCD00143578 CAS Number: 13170-62-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...