N,N-Dibutyl-3-aminophenol ≥95.0%
Supplier: TCI America
D2138-500G
D2138-25G
TCD2138-500GEA
400.06
CAD
TCD2138-500G
TCD2138-025G
N,N-Dibutyl-3-aminophenol ≥95.0%
N,N-Dibutyl-3-aminophenol
CAS Number: 43141-69-1
MDL Number: MFCD00134668
Molecular Formula: C14H23NO
Molecular Weight: 221.34
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Specific Gravity (20/20): 0.98
Formula:
C₁₄H₂₃NO Boiling Pt: 176 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD00134668 CAS Number: 43141-69-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...