Benzyl viologen dichloride ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1,1'-Dibenzyl-[4,4'-bipyridine]-1,1'-diium chloride, 1,1'-Dibenzyl-4,4'-bipyridinium dichloride
D2137-5G
D2137-1G
TCD2137-25GEA
601.13
CAD
TCD2137-25G
TCD2137-005G
TCD2137-001G
Benzyl viologen dichloride ≥98.0% (by HPLC)
Benzyl viologen dichloride
CAS Number: 1102-19-8
MDL Number: MFCD00011991
Molecular Formula: C24H22Cl2N2
Molecular Weight: 409.35
Purity/Analysis Method:
Form: Crystal
Formula:
C₂₄H₂₂Cl₂N₂ MW: 409.35 g/mol |
MDL Number:
MFCD00011991 CAS Number: 1102-19-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...