1,3-Dithiole-2-thione ≥97.0%

Supplier: TCI America

D2133-5G D2133-1G
TCD2133-005GEA 347.17 CAD
TCD2133-005G 76218-996
1,3-Dithiole-2-thione ≥97.0%
1,3-Dithiole-2-thione

CAS Number: 930-35-8
MDL Number: MFCD00014536
Molecular Formula: C3H2S3
Molecular Weight: 134.23
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 110

Formula: C₃H₂S₃
MW: 134.23 g/mol
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00014536
CAS Number: 930-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=002063bc14e7278eb1429d3c72f2b1174ce21e4a&vid=002063bc14e7278eb1429d3c72f2b1174ce21e4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR