Dimethyl-1,3-dithiole-2-thione-4,5-dicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyl 2-Thioxo-1,3-dithiole-4,5-dicarboxylate, 1,3-Dithiole-2-thione-4,5-dicarboxylic Acid Dimethyl Ester
CAS Number: 7396-41-0
MDL Number: MFCD00142532
Molecular Formula: C7H6O4S3
Molecular Weight: 250.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 88
Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Dimethyl 2-Thioxo-1,3-dithiole-4,5-dicarboxylate, 1,3-Dithiole-2-thione-4,5-dicarboxylic Acid Dimethyl Ester
Formula:
C₇H₆O₄S₃ MW: 250.30 g/mol Melting Pt: 88 °C |
MDL Number:
MFCD00142532 CAS Number: 7396-41-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...