4,4'-Dihydroxydiphenylether ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Oxydiphenol, 4,4-Oxydiphenol
D2121-25G
D2121-500G
TCD2121-025GEA
165.43
CAD
TCD2121-025G
TCD2121-500G
4,4'-Dihydroxydiphenylether ≥98.0%
4,4'-Dihydroxydiphenylether
CAS Number: 1965-09-9
MDL Number: MFCD00016463
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168
Product Categories: Materials Chemistry, Monomers, Diphenyl Ethers (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Bis(4-hydroxyphenyl) Ether, 4,4'-Oxydiphenol
Formula:
C₁₂H₁₀O₃ Melting Pt: 168 °C |
MDL Number:
MFCD00016463 CAS Number: 1965-09-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...